1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-Difluoroethanone ≥95%

Supplier: AOBChem USA

340142-1G 340142-500MG 340142-5G 340142-25G
77469-730EA 415.09 USD
77469-730 77482-400 77492-820 77502-658
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-Difluoroethanone ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)-2,2-Difluoroethanone

Precaución: For research use only

Formula: C₉H₃F₇O
CAS Number: 3002460-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR