(2,3-Difluoro-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

340138-1G 340138-500MG 340138-5G 340138-25G
77469-726EA 415.09 USD
77469-726 77482-396 77492-816 77502-654
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(phenyl)methanol ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₄H₉F₅O
CAS Number: 3002453-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR