6-Bromo-2,4-dichloro-5-methylaniline ≥95%

Supplier: AOBChem USA

34010-1G 34010-500MG 34010-5G 34010-25G
77469-718EA 104.03 USD
77469-718 77482-388 77492-806 77502-644
6-Bromo-2,4-dichloro-5-methylaniline ≥95%
6-Bromo-2,4-dichloro-5-methylaniline

Precaución: For research use only

Formula: C₇H₆BrCl₂N
MDL Number: MFCD31619452
CAS Number: 2221812-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR