2',5'-Dimethoxypropiophenone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(2,5-Dimethoxyphenyl)-1-propanone

339207-5G 339207-25G
77492-798EA 460.41 USD
77492-798 77502-634
2',5'-Dimethoxypropiophenone ≥95%
2',5'-Dimethoxypropiophenone

Precaución: For research use only

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00027013
CAS Number: 5803-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR