2-(2-Bromo-3-fluoropyridin-4-yl)butan-2-ol ≥95%

Supplier: AOBChem USA

338733-1G 338733-500MG 338733-5G 338733-25G
77469-690EA 415.09 USD
77469-690 77482-362 77492-778 77502-614
2-(2-Bromo-3-fluoropyridin-4-yl)butan-2-ol ≥95%
2-(2-Bromo-3-fluoropyridin-4-yl)butan-2-ol

Precaución: For research use only

Formula: C₉H₁₁BrFNO
CAS Number: 3002430-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR