Cyclopropyl(2-fluoro-5-methyl-3-(methylthio)phenyl)methanol ≥95%

Supplier: AOBChem USA

338620-1G 338620-500MG 338620-5G 338620-25G
77469-678EA 415.09 USD
77469-678 77482-350 77492-766 77502-602
Cyclopropyl(2-fluoro-5-methyl-3-(methylthio)phenyl)methanol ≥95%
Cyclopropyl(2-fluoro-5-methyl-3-(methylthio)phenyl)methanol

Precaución: For research use only

Formula: C₁₂H₁₅FOS
CAS Number: 3002527-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR