(3,5-Di-tert-butylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

338570-1G 338570-500MG 338570-5G 338570-25G
77469-666EA 415.09 USD
77469-666 77482-338 77492-754 77502-590
(3,5-Di-tert-butylphenyl)(ethyl)sulfane ≥95%
(3,5-Di-tert-butylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₆H₂₆S
CAS Number: 3002535-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR