2-((3-chlorobenzyl)oxy)-3,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

338417-1G 338417-500MG 338417-5G 338417-25G
77469-662EA 415.09 USD
77469-662 77482-334 77492-750 77502-586
2-((3-chlorobenzyl)oxy)-3,5-dimethylbenzaldehyde ≥95%
2-((3-chlorobenzyl)oxy)-3,5-dimethylbenzaldehyde

Precaución: For research use only

Formula: C₁₆H₁₅ClO₂
CAS Number: 3002540-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR