2,5-Dichloro-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

33570-1G 33570-500MG 33570-5G 33570-25G
77469-638EA 253.38 USD
77469-638 77482-312 77492-724 77502-564
2,5-Dichloro-3-fluorobenzaldehyde ≥97%
2,5-Dichloro-3-fluorobenzaldehyde

Precaución: For research use only

Formula: C₇H₃Cl₂FO
MDL Number: MFCD22414486
CAS Number: 1553716-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR