4-Methyl-1H-indazole-5-carbaldehyde ≥95%

Supplier: AOBChem USA

33494-1G 33494-500MG 33494-5G 33494-25G
77469-630EA 224.54 USD
77469-630 77482-304 77492-716 77502-556
4-Methyl-1H-indazole-5-carbaldehyde ≥95%
4-Methyl-1H-indazole-5-carbaldehyde

Precaución: For research use only

Formula: C₉H₈N₂O
MDL Number: MFCD28516467
CAS Number: 1783447-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR