(4-Ethoxyphenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

332838-1G 332838-500MG 332838-5G 332838-25G
77469-594EA 415.09 USD
77469-594 77482-268 77492-680 77502-526
(4-Ethoxyphenyl)(4-ethylphenyl)methanol ≥95%
(4-Ethoxyphenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₇H₂₀O₂
CAS Number: 1284837-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR