Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 4-(4-Methylphenyl)-4-oxobutanoate, 4-Oxo-4-(p-tolyl)butyric acid ethyl ester, Ethyl 4-oxo-4-(p-tolyl)butanoate, 4-(4-Methylphenyl)-4-oxobutanoic acid ethyl ester, Ethyl 4-Oxo-4-(p-tolyl)butyrate

332611-5G 332611-25G
77492-662EA 347.11 USD
77492-662 77502-510
Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95%
Ethyl-4-(4-methylphenyl)-4-oxobutyrate

Precaución: For research use only

Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00266564
CAS Number: 6942-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR