1-(6-Phenylpyridin-3-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 1-(6-Phenyl-3-pyridinyl)ethanone

332008-5G 332008-25G
77492-654EA 475.86 USD
77492-654 77502-504
1-(6-Phenylpyridin-3-yl)ethanone ≥95%
1-(6-Phenylpyridin-3-yl)ethanone

Precaución: For research use only

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12404909
CAS Number: 35022-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR