[2,2'-bithiophen]-5-yltriethylsilane ≥95%
Supplier: AOBChem USA
331441-1G
331441-500MG
331441-5G
331441-25G
77469-558EA
415.09
USD
77469-558
77482-232
77492-640
77502-494
[2,2'-bithiophen]-5-yltriethylsilane ≥95%
[2,2'-bithiophen]-5-yltriethylsilane
Precaución: For research use only
Formula:
C₁₄H₂₀S₂Si |
CAS Number:
2971709-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...