Cyclohexyl(4-isobutoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

331173-1G 331173-500MG 331173-5G 331173-25G
77469-546EA 415.09 USD
77469-546 77482-220 77492-626 77502-480
Cyclohexyl(4-isobutoxyphenyl)methanone ≥95%
Cyclohexyl(4-isobutoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₇H₂₄O₂
CAS Number: 1225559-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR