Ethyl(4-isobutoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

331170-1G 331170-500MG 331170-5G 331170-25G
77469-542EA 415.09 USD
77469-542 77482-216 77492-622 77502-476
Ethyl(4-isobutoxyphenyl)sulfane ≥95%
Ethyl(4-isobutoxyphenyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
CAS Number: 3002423-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR