Cyclopropyl(4-isobutoxyphenyl)methanol ≥95%

Supplier: AOBChem USA

331161-1G 331161-500MG 331161-5G 331161-25G
77469-538EA 415.09 USD
77469-538 77482-212 77492-618 77502-472
Cyclopropyl(4-isobutoxyphenyl)methanol ≥95%
Cyclopropyl(4-isobutoxyphenyl)methanol

Precaución: For research use only

Formula: C₁₄H₂₀O₂
CAS Number: 3002535-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR