5-Bromo-6-chloronicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

33114-5G 33114-25G
77492-610EA 677.74 USD
77492-610 77502-464
5-Bromo-6-chloronicotinaldehyde ≥97%
5-Bromo-6-chloronicotinaldehyde

Precaución: For research use only

Formula: C₆H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD11845760
CAS Number: 71702-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR