1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

331059-1G 331059-500MG 331059-5G 331059-25G
77469-494EA 415.09 USD
77469-494 77482-170 77492-574 77502-432
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-Ethoxy-5-(trifluoromethyl)phenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₄H₁₉F₃O₂
MW: 276.295 g/mol
CAS Number: 3002535-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR