3-Bromo-4-ethoxypyridine ≥95%

Supplier: AOBChem USA

Warning

33073-5G 33073-25G
77492-562EA 726.15 USD
77492-562 77502-422
3-Bromo-4-ethoxypyridine ≥95%
3-Bromo-4-ethoxypyridine

Precaución: For research use only

Formula: C₇H₈BrNO
MW: 202.049 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234192
CAS Number: 3522-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR