(3-Bromo-6-chloro-2-fluorophenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

330712-1G 330712-500MG 330712-5G 330712-25G
77469-478EA 415.09 USD
77469-478 77482-154 77492-558 77502-420
(3-Bromo-6-chloro-2-fluorophenyl)(4-ethylphenyl)methanol ≥95%
(3-Bromo-6-chloro-2-fluorophenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₃BrClFO
CAS Number: 3002471-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR