(2-(Benzyloxy)-5-methylphenyl)(cyclohexyl)methanone ≥95%

Supplier: AOBChem USA

328840-1G 328840-500MG 328840-5G 328840-25G
77469-466EA 415.09 USD
77469-466 77482-142 77492-546 77502-414
(2-(Benzyloxy)-5-methylphenyl)(cyclohexyl)methanone ≥95%
(2-(Benzyloxy)-5-methylphenyl)(cyclohexyl)methanone

Precaución: For research use only

Formula: C₂₁H₂₄O₂
CAS Number: 3002471-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR