1-(2-Ethoxy-5-methylphenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

328630-1G 328630-500MG 328630-5G 328630-25G
77469-458EA 415.09 USD
77469-458 77482-134 77492-538 77502-406
1-(2-Ethoxy-5-methylphenyl)prop-2-en-1-ol ≥95%
1-(2-Ethoxy-5-methylphenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆O₂
CAS Number: 3002422-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR