1-(2-Ethoxy-5-methylphenyl)-2-ethylbutan-1-ol ≥95%

Supplier: AOBChem USA

328628-1G 328628-500MG 328628-5G 328628-25G
77469-454EA 415.09 USD
77469-454 77482-130 77492-534 77502-402
1-(2-Ethoxy-5-methylphenyl)-2-ethylbutan-1-ol ≥95%
1-(2-Ethoxy-5-methylphenyl)-2-ethylbutan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
CAS Number: 1270789-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR