(2-Ethoxy-5-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

328626-1G 328626-500MG 328626-5G 328626-25G
77469-450EA 415.09 USD
77469-450 77482-126 77492-530 77502-398
(2-Ethoxy-5-methylphenyl)(ethyl)sulfane ≥95%
(2-Ethoxy-5-methylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₁H₁₆OS
CAS Number: 3002455-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR