(2-Ethoxy-5-methylphenyl)(propyl)sulfane ≥95%

Supplier: AOBChem USA

328622-1G 328622-500MG 328622-5G 328622-25G
77469-446EA 415.09 USD
77469-446 77482-122 77492-526 77502-394
(2-Ethoxy-5-methylphenyl)(propyl)sulfane ≥95%
(2-Ethoxy-5-methylphenyl)(propyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
CAS Number: 3002442-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR