4-(4-Methoxyphenyl)-2-methyl-3-butyn-2-ol ≥95%

Supplier: AOBChem USA

328158-1G 328158-500MG 328158-5G
77469-438EA 526.33 USD
77469-438 77482-114 77492-518
4-(4-Methoxyphenyl)-2-methyl-3-butyn-2-ol ≥95%
4-(4-Methoxyphenyl)-2-methyl-3-butyn-2-ol

Precaución: For research use only

Formula: C₁₂H₁₄O₂
CAS Number: 84384-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR