2,4'-Dimethylbenzophenone ≥95%

Supplier: AOBChem USA

327700-1G 327700-500MG 327700-5G 327700-25G
77469-426EA 231.75 USD
77469-426 77482-102 77492-506 77502-382
2,4'-Dimethylbenzophenone ≥95%
2,4'-Dimethylbenzophenone

Precaución: For research use only

Formula: C₁₅H₁₄O
CAS Number: 1140-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR