3-(3,5-dibromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥95%

Supplier: AOBChem USA

327232-1G 327232-500MG 327232-5G 327232-25G
77469-422EA 415.09 USD
77469-422 77482-098 77492-502 77502-378
3-(3,5-dibromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol ≥95%
3-(3,5-dibromo-2-fluorophenyl)-2,4-dimethylpentan-3-ol

Precaución: For research use only

Formula: C₁₃H₁₇Br₂FO
CAS Number: 3002441-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR