1-(3,5-dibromo-2-fluorophenyl)propan-1-one ≥95%

Supplier: AOBChem USA

327213-1G 327213-500MG 327213-5G 327213-25G
77469-418EA 415.09 USD
77469-418 77482-094 77492-498 77502-374
1-(3,5-dibromo-2-fluorophenyl)propan-1-one ≥95%
1-(3,5-dibromo-2-fluorophenyl)propan-1-one

Precaución: For research use only

Formula: C₉H₇Br₂FO
CAS Number: 3002488-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR