1-(3-(Benzyloxy)-2,6-dichlorophenyl)prop-2-en-1-ol ≥95%

Supplier: AOBChem USA

327144-1G 327144-500MG 327144-5G 327144-25G
77469-398EA 415.09 USD
77469-398 77482-074 77492-478 77502-354
1-(3-(Benzyloxy)-2,6-dichlorophenyl)prop-2-en-1-ol ≥95%
1-(3-(Benzyloxy)-2,6-dichlorophenyl)prop-2-en-1-ol

Precaución: For research use only

Formula: C₁₆H₁₄Cl₂O₂
CAS Number: 3002436-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR