1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

327134-1G 327134-500MG 327134-5G 327134-25G
77469-394EA 415.09 USD
77469-394 77482-070 77492-474 77502-350
1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3-(Benzyloxy)-2,6-dichlorophenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₁₅H₉Cl₂F₃O₂
CAS Number: 2383561-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR