(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(m-tolyl)methanol ≥95%

Supplier: AOBChem USA

327122-1G 327122-500MG 327122-5G 327122-25G
77469-382EA 415.09 USD
77469-382 77482-058 77492-462 77502-338
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(m-tolyl)methanol ≥95%
(5-Bromo-2-fluoro-3-(trifluoromethyl)phenyl)(m-tolyl)methanol

Precaución: For research use only

Formula: C₁₅H₁₁BrF₄O
CAS Number: 3002509-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR