2-(Benzyloxy)-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

32674-1G 32674-500MG 32674-5G 32674-25G
77469-358EA 213.21 USD
77469-358 77482-034 77492-438 77502-324
2-(Benzyloxy)-3-methylbenzaldehyde ≥97%
2-(Benzyloxy)-3-methylbenzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MDL Number: MFCD12025074
CAS Number: 52803-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR