(2,3-Difluoro-4-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%

Supplier: AOBChem USA

325571-1G 325571-500MG 325571-5G 325571-25G
77469-350EA 415.09 USD
77469-350 77482-026 77492-430 77502-318
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(furan-2-yl)methanol ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(furan-2-yl)methanol

Precaución: For research use only

Formula: C₁₂H₇F₅O₂
CAS Number: 3002455-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR