(2,3-Difluoro-4-(trifluoromethyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

325560-1G 325560-500MG 325560-5G 325560-25G
77469-338EA 415.09 USD
77469-338 77482-014 77492-418 77502-306
(2,3-Difluoro-4-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₀H₁₁F₅Si
CAS Number: 3002514-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR