2-Bromo-5-fluoro-4-(triethylsilyl)pyridine ≥95%

Supplier: AOBChem USA

32486-1G 32486-500MG 32486-5G 32486-25G
77469-322EA 415.09 USD
77469-322 77481-998 77492-402 77502-288
2-Bromo-5-fluoro-4-(triethylsilyl)pyridine ≥95%
2-Bromo-5-fluoro-4-(triethylsilyl)pyridine

Precaución: For research use only

Formula: C₁₁H₁₇BrFNSi
MDL Number: MFCD30489314
CAS Number: 1387561-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR