1-Bromo-2,4-dimethoxy-5-methylbenzene ≥95%

Supplier: AOBChem USA

32416-1G 32416-500MG 32416-5G
77469-310EA 610.79 USD
77469-310 77481-986 77492-388
1-Bromo-2,4-dimethoxy-5-methylbenzene ≥95%
1-Bromo-2,4-dimethoxy-5-methylbenzene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
MDL Number: MFCD30673783
CAS Number: 19345-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR