4-(Hydroxymethyl)-3-methylphenol ≥95%

Supplier: AOBChem USA

32399-1G 32399-500MG 32399-5G 32399-25G
77469-306EA 82.4 USD
77469-306 77481-982 77492-384 77502-280
4-(Hydroxymethyl)-3-methylphenol ≥95%
4-(Hydroxymethyl)-3-methylphenol

Precaución: For research use only

Formula: C₈H₁₀O₂
MDL Number: MFCD07783704
CAS Number: 22574-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR