Cyclohexyl(4-ethoxyphenyl)methanone ≥95%

Supplier: AOBChem USA

323925-1G 323925-500MG 323925-5G
77469-298EA 594.31 USD
77469-298 77481-974 77492-376
Cyclohexyl(4-ethoxyphenyl)methanone ≥95%
Cyclohexyl(4-ethoxyphenyl)methanone

Precaución: For research use only

Formula: C₁₅H₂₀O₂
CAS Number: 92655-14-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR