Ethyl(perfluorophenyl)sulfane ≥95%

Supplier: AOBChem USA

323792-1G 323792-500MG 323792-5G 323792-25G
77469-262EA 415.09 USD
77469-262 77481-938 77492-340 77502-240
Ethyl(perfluorophenyl)sulfane ≥95%
Ethyl(perfluorophenyl)sulfane

Precaución: For research use only

Formula: C₈H₅F₅S
CAS Number: 33288-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR