propyl 3-hydroxy-5-methylhexanoate ≥95%

Supplier: AOBChem USA

323780-1G 323780-500MG 323780-5G 323780-25G
77469-250EA 415.09 USD
77469-250 77481-926 77492-328 77502-228
propyl 3-hydroxy-5-methylhexanoate ≥95%
propyl 3-hydroxy-5-methylhexanoate

Precaución: For research use only

Formula: C₁₀H₂₀O₃
CAS Number: 3002540-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR