5-Bromo-2,4-dimethoxyphenol ≥95%

Supplier: AOBChem USA

32329-1G 32329-500MG 32329-5G
77469-222EA 982.62 USD
77469-222 77481-898 77492-300
5-Bromo-2,4-dimethoxyphenol ≥95%
5-Bromo-2,4-dimethoxyphenol

Precaución: For research use only

Formula: C₈H₉BrO₃
MDL Number: MFCD14534269
CAS Number: 154377-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR