6-Bromo-2-fluoro-3-hydroxybenzonitrile ≥95%

Supplier: AOBChem USA

322201-1G 322201-500MG 322201-5G
77469-202EA 503.67 USD
77469-202 77481-878 77492-280
6-Bromo-2-fluoro-3-hydroxybenzonitrile ≥95%
6-Bromo-2-fluoro-3-hydroxybenzonitrile

Precaución: For research use only

Formula: C₇H₃BrFNO
CAS Number: 1252988-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR