Methyl 4-chloro-2,3-difluorobenzoate ≥97%

Supplier: AOBChem USA

32175-1G 32175-500MG 32175-5G 32175-25G
77469-194EA 125.66 USD
77469-194 77481-870 77492-270 77502-182
Methyl 4-chloro-2,3-difluorobenzoate ≥97%
Methyl 4-chloro-2,3-difluorobenzoate

Precaución: For research use only

Formula: C₈H₅ClF₂O₂
MDL Number: MFCD14698003
CAS Number: 1214334-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR