4-(4-Methylphenyl)-3-butyn-2-one ≥95%

Supplier: AOBChem USA

321114-1G 321114-500MG 321114-5G 321114-25G
77469-186EA 418.18 USD
77469-186 77481-862 77492-262 77502-176
4-(4-Methylphenyl)-3-butyn-2-one ≥95%
4-(4-Methylphenyl)-3-butyn-2-one

Precaución: For research use only

Formula: C₁₁H₁₀O
MW: 158.2 g/mol
CAS Number: 80221-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR