2-Formyl-5-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethoxy)benzaldehyde

32037-1G 32037-500MG 32037-5G
77469-162EA 455.26 USD
77469-162 77481-838 77492-238
2-Formyl-5-(trifluoromethoxy)phenylboronic acid pinacol ester ≥97%
2-Formyl-5-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: For research use only

Formula: C₁₄H₁₆BF₃O₄
Storage Temperature: Ambient
MDL Number: MFCD30344679
CAS Number: 2121513-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR