1-(4-propylphenyl)ethanol ≥95%

Supplier: AOBChem USA

319967-1G 319967-500MG 319967-5G
77469-154EA 597.4 USD
77469-154 77481-830 77492-228
1-(4-propylphenyl)ethanol ≥95%
1-(4-propylphenyl)ethanol

Precaución: For research use only

Formula: C₁₁H₁₆O
CAS Number: 152336-37-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR