2-(isopropylthio)dibenzo[b,d]furan ≥95%

Supplier: AOBChem USA

319827-1G 319827-500MG 319827-5G 319827-25G
77469-118EA 415.09 USD
77469-118 77481-794 77492-192 77502-124
2-(isopropylthio)dibenzo[b,d]furan ≥95%
2-(isopropylthio)dibenzo[b,d]furan

Precaución: For research use only

Formula: C₁₅H₁₄OS
CAS Number: 3002501-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR