Methyl(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)sulfane ≥95%

Supplier: AOBChem USA

319804-1G 319804-500MG 319804-5G 319804-25G
77469-098EA 415.09 USD
77469-098 77481-774 77492-172 77502-104
Methyl(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)sulfane ≥95%
Methyl(2-Methyl-4-((4-Methylbenzyl)oxy)phenyl)sulfane

Precaución: For research use only

Formula: C₁₆H₁₈OS
CAS Number: 3002453-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR